KEY007387558 Key Organics / BIONET

3-amino-3-(4-ethoxy-3-methoxyphenyl)propanoic acid

  • MDL number MFCD02140239
  • Cas Number : 54503-18-3
  • Molecular Weight : 239.271

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 232 - 235
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73494

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3W-0831_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart