KEY007387561 Key Organics / BIONET

5-(5-chlorothiophen-2-yl)-1H-pyrazole

  • MDL number MFCD03791215
  • Cas Number : 166196-61-8
  • Molecular Weight : 184.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168 - 169
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73527

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3W-0877_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart