KEY007387563 Key Organics / BIONET

5-bromo-2-chloro-4,6-dimethylpyridine-3-carbonitrile

  • MDL number MFCD03791216
  • Cas Number : 42951-71-3
  • Molecular Weight : 245.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 106 - 108
Iupac Name 5-bromo-2-chloro-4,6-dimethylnicotinonitrile
Inchi Code 1S/C8H6BrClN2/c1-4-6(3-11)8(10)12-5(2)7(4)9/h1-2H3
Inchi Key UUEREACRVVTQKF-UHFFFAOYSA-N

Description

Melting Point 106 - 108
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73546

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/3W-0900_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart