KEY007387569 Key Organics / BIONET

N-(2,4-dichlorophenyl)-2-methyloxirane-2-carboxamide

  • MDL number MFCD01935966
  • Cas Number : 65925-78-2
  • Molecular Weight : 246.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51 - 59
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75035

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4J-585S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart