KEY007387577 Key Organics / BIONET

methyl (2E)-3-[(E)-4-nitrobenzoyloxy]prop-2-enoate

  • MDL number MFCD03001264
  • Cas Number : 1104461-74-6
  • Molecular Weight : 251.194

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152 - 154
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76448

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4R-0243_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart