KEY007387578 Key Organics / BIONET

1-(4-nitrobenzenesulfonyl)piperidine-4-carboxylic acid

  • MDL number MFCD01045658
  • Cas Number : 331828-49-0
  • Molecular Weight : 314.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 248 - 250
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76723

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4R-1331_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart