KEY007387579 Key Organics / BIONET

2-(5-nitro-2-oxo-2,3-dihydro-1,3-benzoxazol-3-yl)acetamide

  • MDL number MFCD03425771
  • Cas Number : 439095-26-8
  • Molecular Weight : 237.171

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 234 - 235
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76771

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4T-0026_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart