KEY007387580 Key Organics / BIONET

methyl 4-(3-amino-4-nitrophenoxy)benzoate

  • MDL number MFCD03425772
  • Cas Number : 439095-28-0
  • Molecular Weight : 288.259

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76789

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4T-0210_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart