KEY007387582 Key Organics / BIONET

3-{7-thia-2,5-diazatricyclo[6.4.0.0^{2,6}]dodeca-1(8),3,5,9,11-pentaen-4-yl}aniline

  • MDL number MFCD03791217
  • Cas Number : 79890-07-6
  • Molecular Weight : 265.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169 - 171
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76918

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4T-0640_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart