KEY007387588 Key Organics / BIONET

1-methyl-4-(5-nitropyridin-2-yl)piperazine

  • MDL number MFCD03001270
  • Cas Number : 55403-34-4
  • Molecular Weight : 222.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 108 - 110
Iupac Name 1-methyl-4-(5-nitro-2-pyridinyl)piperazine
Inchi Code 1S/C10H14N4O2/c1-12-4-6-13(7-5-12)10-3-2-9(8-11-10)14(15)16/h2-3,8H,4-7H2,1H3
Inchi Key DZIASOMGGPNYCU-UHFFFAOYSA-N

Description

Melting Point 108 - 110
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80237

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/5R-0318_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart