KEY007387593 Key Organics / BIONET

5-(2,4-dichlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD01935965
  • Cas Number : 321848-48-0
  • Molecular Weight : 339.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127.5 - 129
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82428

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6K-310S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart