KEY007387595 Key Organics / BIONET

[3-(4-methoxyphenyl)-1,2-oxazol-5-yl]methanamine

  • MDL number MFCD01935989
  • Cas Number : 338982-43-7
  • Molecular Weight : 204.229

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119 - 122
Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82885

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/6M-352S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart