KEY007387598 Key Organics / BIONET

11-(2-methylprop-2-enamido)undecanoic acid

  • MDL number MFCD03425798
  • Cas Number : 59178-93-7
  • Molecular Weight : 269.385

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73 - 74
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84040

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6T-0302_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart