KEY007387600 Key Organics / BIONET

ethyl 1-(quinoxalin-6-yl)piperidine-4-carboxylate

  • MDL number MFCD03425801
  • Cas Number : 453557-64-7
  • Molecular Weight : 285.347

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69 - 71
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84054

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6T-0323_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart