KEY007387604 Key Organics / BIONET

1-[2,3-dichloro-4-(ethanesulfonyl)phenyl]ethan-1-one

  • MDL number MFCD01935964
  • Cas Number : 338982-46-0
  • Molecular Weight : 281.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88 - 90
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85684

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7J-348S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart