KEY007387611 Key Organics / BIONET

1,3-dimethyl 2-(4-nitrophenyl)propanedioate

  • MDL number MFCD03425806
  • Cas Number : 4033-88-9
  • Molecular Weight : 253.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 125 - 127
Iupac Name dimethyl 2-(4-nitrophenyl)malonate
Inchi Code 1S/C11H11NO6/c1-17-10(13)9(11(14)18-2)7-3-5-8(6-4-7)12(15)16/h3-6,9H,1-2H3
Inchi Key PYUABAXAUVLRNX-UHFFFAOYSA-N

Description

Melting Point 125 - 127
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87671

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7T-0054_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart