KEY007387619 Key Organics / BIONET

ethyl 6-methyl-4-(4-methylphenyl)-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate

  • MDL number MFCD00719779
  • Cas Number : 299949-24-9
  • Molecular Weight : 274.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218 - 219
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87781

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7T-0311_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart