KEY007387622 Key Organics / BIONET

methyl 3-(5-formylfuran-2-yl)thiophene-2-carboxylate

  • MDL number MFCD00052169
  • Cas Number : 255828-29-6
  • Molecular Weight : 236.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118 - 119
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88528

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8D-127_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart