KEY007387624 Key Organics / BIONET

ethyl 4-(2,5-dimethyl-1H-pyrrol-1-yl)benzoate

  • MDL number MFCD00022462
  • Cas Number : 5159-70-6
  • Molecular Weight : 243.306

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90 - 92
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90446

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8R-0002_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart