KEY007387628 Key Organics / BIONET

5-methoxy-2-(1H-pyrrol-1-yl)aniline

  • MDL number MFCD00047063
  • Cas Number : 59194-26-2
  • Molecular Weight : 188.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 41 - 43
Iupac Name 5-methoxy-2-(1H-pyrrol-1-yl)aniline
Inchi Code 1S/C11H12N2O/c1-14-9-4-5-11(10(12)8-9)13-6-2-3-7-13/h2-8H,12H2,1H3
Inchi Key LKSVZBCORDEARF-UHFFFAOYSA-N

Description

Melting Point 41 - 43
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90900

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/8T-0200_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart