KEY007387632 Key Organics / BIONET

5-acetyl-6-methyl-2-oxo-1,2-dihydropyridine-3-carboxamide

  • MDL number MFCD03425816
  • Cas Number : 52600-60-9
  • Molecular Weight : 194.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 325 - 327
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90912

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8T-0220_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart