KEY007387633 Key Organics / BIONET

1-(cyclooctylamino)-6-methyl-9H-xanthen-9-one

  • MDL number MFCD03425817
  • Cas Number : 886360-53-8
  • Molecular Weight : 335.447

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126 - 129
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90920

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8T-0236_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart