KEY007387635 Key Organics / BIONET

ethyl 5-carbamoyl-4-methyl-6-sulfanylidene-1,6-dihydropyridine-3-carboxylate

  • MDL number MFCD03425818
  • Cas Number : 171113-26-1
  • Molecular Weight : 240.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 271 - 273
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90941

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8T-0270_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart