KEY007387636 Key Organics / BIONET

3,8-dioxo-2,3,5,6,7,8-hexahydroisoquinoline-4-carboxamide

  • MDL number MFCD03425819
  • Cas Number : 171113-31-8
  • Molecular Weight : 206.201

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 315 - 317
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/90942

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8T-0271_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart