KEY007387643 Key Organics / BIONET

2-amino-1-benzyl-4,5-diphenyl-1H-pyrrole-3-carbonitrile

  • MDL number MFCD01935933
  • Cas Number : 55817-67-9
  • Molecular Weight : 349.437

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175 - 179
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/92651

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9L-336S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart