KEY007387650 Key Organics / BIONET

1,3-dimethyl 2-(5-nitropyridin-2-yl)propanedioate

  • MDL number MFCD03425824
  • Cas Number : 68719-87-9
  • Molecular Weight : 254.198

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103.5
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93869

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9T-0002_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart