KEY007387662 Key Organics / BIONET

ethyl 1-(6-chloropyridazin-3-yl)piperidine-4-carboxylate

  • MDL number MFCD03425828
  • Cas Number : 252263-47-1
  • Molecular Weight : 269.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55 - 60
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93936

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9T-0248_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart