KEY007387663 Key Organics / BIONET

4-[(3E)-3-(hydroxyimino)-2-[(1E)-1-(hydroxyimino)ethyl]butyl]phenol

  • MDL number MFCD03425829
  • Cas Number : 454473-86-0
  • Molecular Weight : 236.271

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210 - 212
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93960

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9T-0292_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart