KEY007387664 Key Organics / BIONET

(2E)-3-chloro-2,3-bis(4-methoxyphenyl)prop-2-enal

  • MDL number MFCD00547134
  • Cas Number : 181308-92-9
  • Molecular Weight : 302.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160 - 163
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93977

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9T-0314_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart