KEY013018418 Key Organics / BIONET

1,3-dimethyl 2-[2,6-dinitro-4-(trifluoromethyl)phenyl]propanedioate

  • MDL number MFCD03787071
  • Cas Number : 865658-18-0
  • Molecular Weight : 366.205

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101 - 103
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73347

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3W-0036_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart