KEY013018455 Key Organics / BIONET

3-(1H-pyrrol-1-yl)-1-benzofuran-2-carboxylic acid

  • MDL number MFCD00445255
  • Cas Number : 80066-99-5
  • Molecular Weight : 227.219

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 242 - 244
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73402

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3W-0301_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart