KEY013018540 Key Organics / BIONET

(2E)-3-(dimethylamino)-1-(pyrazin-2-yl)prop-2-en-1-one

  • MDL number MFCD02325230
  • Cas Number : 111781-53-4
  • Molecular Weight : 177.207

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131 - 133
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73524

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3W-0873_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart