KEY013018649 Key Organics / BIONET

5-amino-1-(3-nitropyridin-2-yl)-1H-pyrazole-4-carbonitrile

  • MDL number MFCD03787714
  • Molecular Weight : 230.187

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215 - 217
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77126

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4W-0330_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart