KEY013018651 Key Organics / BIONET

3-{3-[3-(trifluoromethyl)phenyl]-1,2,4-oxadiazol-5-yl}propanoic acid

  • MDL number MFCD03787716
  • Cas Number : 866018-88-4
  • Molecular Weight : 286.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90 - 92
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77128

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4W-0335_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart