KEY013018697 Key Organics / BIONET

5-bromo-4,6-dimethyl-1H-pyrazolo[3,4-b]pyridin-3-amine

  • MDL number MFCD03541051
  • Cas Number : 42951-65-5
  • Molecular Weight : 241.092

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 279
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77181

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4W-0834_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart