KEY013018715 Key Organics / BIONET

3-(2,5-dimethyl-1H-pyrrol-1-yl)-3-(thiophen-3-yl)propanoic acid

  • MDL number MFCD03848568
  • Cas Number : 866019-28-5
  • Molecular Weight : 249.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112 -115
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77200

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4W-0858_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart