KEY013018759 Key Organics / BIONET

1,3-dimethyl 2-(2-chloro-6-nitrophenyl)propanedioate

  • MDL number MFCD03787565
  • Cas Number : 147124-36-5
  • Molecular Weight : 287.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132 - 134
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80762

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5W-0074_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart