KEY013018760 Key Organics / BIONET

1,3-dimethyl 2-(4-chloro-2-nitrophenyl)propanedioate

  • MDL number MFCD00179451
  • Cas Number : 147124-32-1
  • Molecular Weight : 287.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 83 - 85
Iupac Name dimethyl 2-(4-chloro-2-nitrophenyl)malonate
Inchi Code 1S/C11H10ClNO6/c1-18-10(14)9(11(15)19-2)7-4-3-6(12)5-8(7)13(16)17/h3-5,9H,1-2H3
Inchi Key XNQGJNNDVKLNQO-UHFFFAOYSA-N

Description

Melting Point 83 - 85
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80763

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/5W-0075_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart