KEY013018945 Key Organics / BIONET

1-(4-chloro-3-nitrobenzenesulfonyl)piperidine-4-carboxylic acid

  • MDL number MFCD03787468
  • Cas Number : 368427-83-2
  • Molecular Weight : 348.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104 - 142
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84161

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6W-0230_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart