KEY013019013 Key Organics / BIONET

1-(benzylsulfanyl)-2-nitrobenzene

  • MDL number MFCD00031120
  • Cas Number : 22057-44-9
  • Molecular Weight : 245.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80 - 81
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87837

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7W-0001_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart