KEY013019017 Key Organics / BIONET

1-{[(2-chlorophenyl)methyl]sulfanyl}-4-nitrobenzene

  • MDL number MFCD03341001
  • Cas Number : 866132-59-4
  • Molecular Weight : 279.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 119 - 120
Iupac Name 1-chloro-2-{[(4-nitrophenyl)sulfanyl]methyl}benzene
Inchi Code 1S/C13H10ClNO2S/c14-13-4-2-1-3-10(13)9-18-12-7-5-11(6-8-12)15(16)17/h1-8H,9H2
Inchi Key HMJLAFGWFMNEPJ-UHFFFAOYSA-N

Description

Melting Point 119 - 120
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87841

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7W-0016_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart