KEY013019153 Key Organics / BIONET

3-fluoro-2-methyl-6-nitroaniline

  • MDL number MFCD03787364
  • Cas Number : 485832-96-0
  • Molecular Weight : 170.143

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118 - 121
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93717

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9R-0643_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart