KEY014415594 Key Organics / BIONET

1,2,3,4,5,6,7,8-octahydroquinoline-2,5-dione

  • MDL number MFCD01111799
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 196 - 198
Iupac Name 4,6,7,8-tetrahydro-2,5(1H,3H)-quinolinedione
Inchi Code 1S/C9H11NO2/c11-8-3-1-2-7-6(8)4-5-9(12)10-7/h1-5H2,(H,10,12)
Inchi Key KNRFNTVMEIIKEB-UHFFFAOYSA-N

Description

Melting Point 196 - 198
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88021

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7W-0869_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart