KEY017860806 Key Organics / BIONET

(2Z)-2-(4-chlorophenyl)-3-(dimethylamino)prop-2-enal

  • MDL number MFCD04973720
  • Cas Number : 19927-64-1
  • Molecular Weight : 209.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123 - 125
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71314

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3J-553S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart