KEY017860808 Key Organics / BIONET

1-methyl-3-(trifluoromethyl)-1H-thieno[2,3-c]pyrazole-5-carbohydrazide

  • MDL number MFCD04973722
  • Cas Number : 886360-63-0
  • Molecular Weight : 264.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218 - 220
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73670

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3X-0720_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart