KEY017860814 Key Organics / BIONET

6-(2,6-dimethylmorpholin-4-yl)pyridine-3-carbaldehyde

  • MDL number MFCD04973726
  • Cas Number : 886360-67-4
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp oil
Iupac Name 6-(2,6-dimethyl-4-morpholinyl)nicotinaldehyde
Inchi Code 1S/C12H16N2O2/c1-9-6-14(7-10(2)16-9)12-4-3-11(8-15)5-13-12/h3-5,8-10H,6-7H2,1-2H3
Inchi Key QJKIFMOLBXODKC-UHFFFAOYSA-N

Description

Melting Point oil
Purity 95%
Physical Form Liquid,Low Melting Solid,Gum
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77311

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/4X-0716_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart