KEY017860823 Key Organics / BIONET

1,3-diethyl 2-cyclohexylpropanedioate

  • MDL number MFCD00019363
  • Cas Number : 2163-44-2
  • Molecular Weight : 242.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp oil
Iupac Name diethyl 2-cyclohexylmalonate
Inchi Code 1S/C13H22O4/c1-3-16-12(14)11(13(15)17-4-2)10-8-6-5-7-9-10/h10-11H,3-9H2,1-2H3
Inchi Key FUOPELSDMOAUBM-UHFFFAOYSA-N

Description

Melting Point oil
Purity 95%
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77323

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/4X-0732_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart