KEY017860824 Key Organics / BIONET

(6-chloropyridin-3-yl)methyl acetate

  • MDL number MFCD04973731
  • Cas Number : 881734-66-3
  • Molecular Weight : 185.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92 @ 0.01 mmHg
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77345

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4X-0830_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart