KEY017860825 Key Organics / BIONET

N-(3-methoxyphenyl)-2,2-dimethylpropanamide

  • MDL number MFCD00662896
  • Cas Number : 56619-93-3
  • Molecular Weight : 207.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 125 - 126
Iupac Name N-(3-methoxyphenyl)-2,2-dimethylpropanamide
Inchi Code 1S/C12H17NO2/c1-12(2,3)11(14)13-9-6-5-7-10(8-9)15-4/h5-8H,1-4H3,(H,13,14)
Inchi Key DAFHCFQPQMYDFI-UHFFFAOYSA-N

Description

Melting Point 125 - 126
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81013

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/5X-0602_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart