KEY017861154 Key Organics / BIONET

1-tert-butyl-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD04124379
  • Cas Number : 142818-02-8
  • Molecular Weight : 236.194

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115 - 117
Purity 95
Storage temp. RT
Physical Form White to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7144

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11W-0279_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart